Letku, kumia, Versilon™ GA
Toimittaja: Saint Gobain Life Sciences
702525
228-1374EA 166 EUR
228-1374
Letku, kumia, Versilon™ GA
Letku Siirtoputki
Natural rubber, light brown, slightly translucent.
  • Excellent elasticity, 500% extension possible
  • Suitable for steam sterilisation
  • Complies with the requirements of the US Food and Drugs Agency FDA
This tubing offers excellent elasticity and resilience. It is easy to fit on all types of connectors; it has similar properties to latex and replaces it in most laboratory and general industrial applications. Typical applications include general laboratory, blood and tissue evaluation, all kinds of dispensing, soap and cleaners, alcohol and dairy.