Hanat, letkuventtiilit
Toimittaja: KARTELL
371 374 376 379
227-4010EA 4.3 EUR
227-4010 227-4008 227-4013 227-4015
Hanat, letkuventtiilit
Hanat
HDPE with PP tap.
  • Good chemical resistance
  • No lubricants required
  • Good thermal resistance
These flow through valves are ideal for regulating or stopping the flow of gases and liquids in tube connections under low pressure (up to 0,5 bar) or partial vacuum. Smooth connectors, slightly tapered on both sides.