Hanat, palloventtiilit, tynnyreille, LaboPlast®
Toimittaja: BURKLE
0500-8003 0500-8001 0500-0134 0500-8002
227-1108EA 104 EUR
227-1108 227-1109 BURK0500-0134 BURK0500-8002
Hanat, palloventtiilit, tynnyreille, LaboPlast®
Hanat
PP, black/red
  • Suitable for solidifying media
  • Max. 4 bar pressure at 20 °C
  • Individually adaptable, through various accessories
Ball valves for barrels with thread connections.