Indium arsenide ≥99% (metals basis)

Toimittaja: Thermo Fisher Scientific
Vaara

22651.04 22651.09
22651.04EA 225 EUR
22651.04 22651.09
Indium arsenide ≥99% (metals basis)
Indium arsenide

Purity: 99% CAS No.: 1303-11-3 Molecular Formula: AsIn Formula Weight: 189.74 Physical Form: 5mm & Down Pieces Odour: Odourless Merck Reference: 14,4949 UN No.: 1557 MDL No.: MFCD00016144


In semiconductor electronics

Kaava: AsIn
Sulamispiste: 936 °C
Tiheys: 5,67 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00016144
CAS-numero: 1303-11-3
EINECS: 215-115-3
UN: 1557
ADR: 6.1,III
Merck Index: 14,4949

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR