Setrimonibromidi, heksadekyylitrimetyyliammoniumbromidi ≥99.0%, AnalaR NORMAPUR® analytical reagent

Toimittaja: VWR Chemicals
Vaara

Synonyms: N-Cetyl-N,N,N-trimethylammonium bromide, Cetyltrimethylammonium bromide, Hexadecyltrimethylammonium bromide, CTAB, (1-Hexadecyl)trimethylammonium bromide, N,N,N-Trimethylhexadecan-1-aminium bromide

22610.132EA 29.6 EUR
22610.132 22610.180 22610.290 22610.460
Setrimonibromidi, heksadekyylitrimetyyliammoniumbromidi ≥99.0%, AnalaR NORMAPUR® analytical reagent
Setrimonibromidi, heksadekyylitrimetyyliammoniumbromidi
Kaava: CH₃(CH₂)₁₅N(Br)(CH₃)₃
MW: 364.45 g/mol
Sulamispiste: 237…243 °C
Leimahduspiste: 244 °C (sulj. astia)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011772
CAS-numero: 57-09-0
EINECS: 200-311-3
UN: 3077
ADR: 9 III
Merck Index: 13,02034

Order Now

Testitulosten erittely

Assay Min. 99.0 %
Heavy metals (as Pb) Max. 10 ppm
Ignition residue (SO4) Max. 0.1 %
Water Max. 1 %
Fe (Iron) Max. 10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR