Peristalttiset pumppusetit
Toimittaja: Heidolph Instruments GmbH & Co.KG
523-50019-00 523-50019-02 523-50068-00 523-50068-02 523-50068-03
224-1455EA 4190 EUR
224-1455 224-1456 224-0032 224-0040 HEID523-50068-03
Peristalttiset pumppusetit
Pumput Peristalttiset pumput
Packages for Hei-FLOW Value 01 Hei-FLOW Value 06 peristaltic pumps.
  • For standard applications and liquid transfer
Hei-FLOW Silver 1 Package includes Hei-FLOW Value 01 (model PD 5001), Pump head SP quick 1,6, and 1 m each Tygon® (standard) and Silicone tubing (inner Ø 3,1 mm).
Hei-FLOW Silver 2 Package includes Hei-FLOW Value 06 (model PD 5006), Pump head SP standard 2,5, and 1 m each Tygon® (standard) and Silicone tubing (inner Ø 6,4 mm).
Tilaustiedot: For more information, see peristaltic pump drive models PD 5001 and PD 5006.