Natriumkarbonaatti ≥99.5%, powder ACS, Fluka™

Toimittaja: Honeywell Chemicals
Varoitus

Synonyms: Calcined soda, Carbonic acid disodium salt

223530-1KG 223530-2.5KG 223530-500G
223530-1KGEA 75 EUR
223530-1KG 223530-2.5KG 223530-500G
Natriumkarbonaatti ≥99.5%, powder ACS, Fluka™
Natriumkarbonaatti
Kaava: Na₂CO₃
Kiehumispiste: 1600 °C (1013 hPa)
Sulamispiste: 854 °C
Tiheys: 2,53 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00003494
CAS-numero: 497-19-8
EINECS: 207-838-8
REACH: 01-2119485498-19
Merck Index: 14,08596
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a6ff30f1487cdef86add98b38bfa8eb3dac14f6&vid=c5b129b7acb5c897b9d970ea3130d61fb1934b54&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Identity (IR) complying
Assay (excluding ignited substance) Min. 99.5 %
Loss on ignition (285°C) Max. 1.0 %
Water insoluble matter Max. 0.01 %
Calcium (Ca) Max. 0.03 %
Iron (Fe) Max. 5 ppm
Potassium (K) Max. 0.005 %
Magnesium (Mg) Max. 0.005 %
Heavy metals (ICP) ACS Max. 5 ppm
Silicate (as SiO₂) Max. 0.005 %
Chloride (Cl) Max. 0.001 %
Phosphate (PO₄) Max. 0.001 %
Total S (as SO₄) Max. 0.003 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR