Annostelupumppu, jalkakäyttöinen, OTAL®
Toimittaja: BURKLE
5000-1500 5000-2200 5000-4016 5000-5018 5000-1800 5000-3012 5000-6000
223-0042EA 192 EUR
223-0042 223-1122 223-0004 223-0005 223-1123 223-1124 223-0117
Annostelupumppu, jalkakäyttöinen, OTAL®
Pumput Rumpupumput
The telescopic immersion tube adapts itself to the container height on insertion of the foot pump. This ensures good removal of residual liquid and means that the foot pump can be used with different containers.
  • Simple handling: Both hands remain free for filling
  • Very good pump capacity due to large volume foot bellows
  • With universal bungs made of soft PVC, for container openings of 30 to 70 mm Ø
OTAL® foot pump made of PP
OTAL® foot pumps with delivery tubes made of PP, resistant to weak acids, alkalis and detergents.
OTAL® foot pump made of PVDF