Ammoniakkiliuos 28,0 - 30,0% (NH₃ basis) in water ACS, Fluka™

Toimittaja: Honeywell Chemicals
Vaara

Synonyms: Ammoniakki

221228-1L 221228-500ML 221228-6X500ML 221228-100ML 221228-2.5L
221228-1LEA 53.1 EUR
221228-1L 221228-500ML 221228-6X500ML 221228-100ML 221228-2.5L
Ammoniakkiliuos 28,0 - 30,0% (NH₃ basis) in water ACS, Fluka™
Ammoniakkiliuos
Kaava: NH₃
MW: 35.05 g/mol
Kiehumispiste: 32 °C (1.013 hPa)
Sulamispiste: –92 °C
Tiheys: 0.89 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00066650
CAS-numero: 1336-21-6
UN: 2672
ADR: 8 III

Order Now

Testitulosten erittely

Appearance complying
Assay (acidimetric) 28.0 - 30.0 %
Residue on ignition Max. 0.002 %
Iron (Fe) Max. 0.00002 %
Heavy metals (as Pb) Max. 0.00005 %
KMnO₄ reducing matter complying
CO₂ Max. 0.002 %
Chloride (Cl) Max. 0.00005 %
Nitrate (NO₃) Max. 0.0002 %
Sulfate (SO₄) Max. 0.0002 %
Phosphate (PO₄) Max. 0.0002 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR