Funnels, powder
Toimittaja: Kavalier
632413103050 632413103060 632413103080 632413103120 632413103160 632413103200 632413103100
221-1511EA 29.8 EUR
221-1511 221-1512 221-1514 221-1516 221-1517 221-1518 KAVA632413103100
Funnels, powder
Suppilot Jauhesuppilot
Borosilicate glass 3,3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With smooth internal walls. For transferring powdery materials. Also suitable for use with liquids. Funnel rim is ground and fire-polished.