Stoppers for conical ground joints
Toimittaja: Lenz Laborglas GmbH & CO.KG
01400512 01400529 01400514 01400519
217-9096EA 13.5 EUR
217-9096 217-9099 217-9097 217-9098
Stoppers for conical ground joints
Tulpat Lasi
DURAN®, borosilicate glass stopper, clear, solid or semi hollow.
  • Excellent chemical resistance
  • High thermal resistance and minimal heat expansion results in high resistance to temperature changes
  • Octagonal top allows easy release and removal of the stopper
Sertifikaatit: DIN 12252