Tulpat, silikonia, HelixMark®
Toimittaja: Freudenberg Medical Europe GmbH
70-416-05 70-416-00 70-416-01 70-416-02 70-416-03 70-416-04 70-416-06 70-416-07 70-416-08 70-416-09 70-416-10 70-416-12 70-416-90 70-416-99
217-0654EA 1250 EUR
217-0654 217-0655 217-0656 217-0657 217-0658 217-0659 217-0660 217-0661 217-0662 217-0663 217-0664 217-0665 217-0666 217-0667
Tulpat, silikonia, HelixMark®
Tulpat Silikoni
HelixMark silicone stoppers enable reliable sealing of glass tubes, vials and flasks. The stoppers are suitable for all commonly used diameters and sizes.
  • Stoppers with two holes upon request
Typical Hardness: 50 Shore
ISO 13485
Sertifikaatit: The stoppers meet ISO 10993-1, FDA 21 CFR 177-2600 and USP Class VI requirements, EP: EP 3.1.9
Toimitustiedot: Material certification and batch number included in every box.
Pakkaus: Packaged in boxes with 12 gaskets per resealable bag.