Tulpat, kumia, kartiomallinen
Toimittaja: KARTELL
1136 1138 1144 1145 1147 1148
217-0471EA 1.1 EUR
217-0471 217-0473 217-0478 217-0479 217-0481 217-0482
Tulpat, kumia, kartiomallinen
Tulpat Kumi
Natural red rubber with elastic, tight fitting design.
  • Excellent seal and adhesion
  • Good chemical resistance
  • Non toxic
  • Temperature resistant from −25 to +70 °C