Crucibles, broad base
Toimittaja: TECHNICAL QUARTZ SOLUTIONS
73081 73082 73083
216-4496EA 155 EUR
216-4496 216-4497 216-4498
Crucibles, broad base
Upokkaat
These crucibles are made from fused quartz, translucent, glazed.
  • Max. temperature: 1050 °C
Toimitustiedot: Supplied without lid.