Cylinders, without lid
Toimittaja: GLASWARENFABRIK KARL HECHT
44405012 44405018 44405054 44405024 44405066 44405072 44405078 44405042 44409024 44409036 44411036 44409030 44411024 44405036 44440014 44411030 44409018 44440028
HECH44405012EA 12.2 EUR
HECH44405012 HECH44405018 HECH44405054 216-4289 HECH44405066 216-4290 216-4291 216-4292 HECH44409024 HECH44409036 HECH44411036 HECH44409030 HECH44411024 HECH44405036 HECH44440014 HECH44411030 HECH44409018 HECH44440028
Cylinders, without lid
Mittalasit Säilytyssylinterit
Jars, made of glass or stainless steel, with base, without lids.
Suitable for storing dressing material, cotton, clinical thermometers, forceps and other instruments.
Tilaustiedot: Lids with knob available must be ordered separately.
Order Now
TEKNISET TIEDOT
Environmentally Preferable