VWR®, Perforated Euro Containers
Toimittaja: VWR Collection
216-2418EA 10.6 EUR
216-2418 216-2419 216-2427 216-2428 216-2429 216-2432 216-2433 216-2434 216-2435 216-2436 216-2438 216-2437
VWR®, Perforated Euro Containers
Laatikot Kuljetuslaatikot
These perforated plastic stackable Euro containers are made from heavy-duty polypropylene, suitable for every industry and for any application. They provide outstanding air and liquid permeability.
  • Made of PPC (polypropylene copolymer)
  • Optimal air circulation
  • Hygienically flawless and easy to clean
  • Ergonomic handles for effective manual handling
  • Suitable for –20 to +65 °C storage
  • 100% recyclable
The perforated Euro containers are available in universal Euros sizes: 400×300, 600×400 or 800×600 mm. They are perfect for palletisation. They are suitableMore Product Information