Laboratory trays, PP
Toimittaja: MULTIROIR
1085 1086 1087 1088 1187-12
216-2239EA 5.3 EUR
216-2239 216-2240 216-2241 216-2242 MULC1187-12
Laboratory trays, PP
Haihdutusmaljat/Tarjottimet/Altaat Laboratorioalustat/-astiat
Range of five lightweight and sturdy laboratory trays. They are made of white heavy-duty PP and are ideal for a wide range of uses.
  • Food grade
  • Nestable
  • Thermal resistance –25 to +70 °C
  • Machine washable
Smooth easy to clean hygienic internal surface.