Containers, wide mouth, with screw cap, PACKO
Toimittaja: CURTEC
4315-50-048 4320-50-048 4325-50-048 4305-57-048 4303-57-048 4306-57-048 4310-57-048 4313-57-048
216-1994EA 57.3 EUR
216-1994 216-1995 216-1996 216-1991 216-1990 216-1838 216-1992 216-1993
Containers, wide mouth, with screw cap, PACKO
Näyteastiat, muovia Yleiskäyttöinenastiat
Small volume white HDPE container with dark red screw lid and translucent innerseal. For storage and sample shipment of powders, pastes and liquids. Easy to close and open with just a quarter-turn.
  • Water vapour tight
  • Tamper evident
  • UN marking (1H2) for hazardous solids
  • Smooth inside
  • Food grade
Protect your valuable products against moisture, tampering and contamination. Reduce handling time. Make sure your hazardous solids arrive safely at their destination.