Transport and Storage Container with 'Activ Lock' System
Toimittaja: MULTIROIR
6411 6416 6461
216-1985EA 30.8 EUR
216-1985 216-1986 216-1987
Transport and Storage Container with 'Activ Lock' System
Laatikot Säilytyslaatikot
Foldable containers made of black PP with 'active lock' system for fast, easy and safe operation. These folding containers are light weight and very easy to open and fold down.
  • Folds easily without applying any pressure
  • 'Active lock' secures the RPC when erected
  • Panels fold easily and quietly when lock is released
  • Foldable and stackable to maximize truck and store space
  • Support cross stacking
  • Extra lightweight for handling ease
  • Easy to clean polymer resists moisture, insects and fungi; impervious to acids, fats, solvents and odors
Long-lasting cratesMore Product Information