Dishes, crystallising
Toimittaja: SciLabware
1470/02D 1470/04D 1470/08D 1470/10D 1470/12D 1470/16D 1471/12M 1470/01M 1471/00M 1471/02M 1471/08M 1471/10M 1471/16M 1470/03M 1471/01M 1471/03M 1471/04M
216-2762EA 170 EUR
216-2762 216-2763 216-2764 216-2765 216-2766 216-2767 216-1551 216-1400 216-1544 216-1546 216-1549 216-1550 216-1552 BIBB1470/03M BIBB1471/01M BIBB1471/03M BIBB1471/04M
Dishes, crystallising
Haihdutusmaljat/Tarjottimet/Altaat Kiteytysmaljat
PYREX®, borosilicate glass, with or without spout.
  • Very good chemical resistance
  • High temperature resistance up to +500 °C
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Features flat base and straight sides and white labelling area.