Instrument dishes
Toimittaja: GLASWARENFABRIK KARL HECHT
42900001 42900010 42900002 42900003 42905002 42905003 42905004 42905005 42905007 42905001
216-1401EA 0 EUR
216-1401 216-1408 216-1402 216-1403 216-1412 216-1413 216-1414 216-1415 216-1417 216-1411
Instrument dishes
Haihdutusmaljat/Tarjottimet/Altaat Instrumenttialustat/-astiat
Thick-walled glass.
  • Can be sterilised in hot air up to 180 °C
  • Robust
  • Stainless steel lids are available separately
Multipurpose dishes with vertical walls for a variety of applications in the laboratory, field use and industry.