Laboratoriotarjottimet/ Altaat, LaboPlast®
Toimittaja: BURKLE
4201-1824 4201-2430 4201-3040 4201-1318 4201-4050 4201-5070 4201-0095 4201-0100
216-1342EA 26.9 EUR
216-1342 216-1343 216-1344 216-1341 216-1345 216-1346 216-0899 216-0922
Laboratoriotarjottimet/ Altaat, LaboPlast®
Haihdutusmaljat/Tarjottimet/Altaat Laboratorioalustat/-astiat
PP, valkoinen.
  • Manufactured using the injection moulding process, not by deep drawing
  • Trays can be nested
  • Trays not suitable for temperature controlled cupboards or dishwashers
  • Temperature resistent up to 60 °C (briefly up to 80 °C)
  • With EU foodstuffs approval
The laboratory trays are practical all-purpose trays that can be nested when empty, also durable, shatterproof and resist flexing. The trays have a strong, rounded rim. Suitable as a spill trough for environmentally dangerous, aggressive substances, for experiments, as an etching tray, as anMore Product Information