Dishes, evaporating
Toimittaja: QUARTZTECHNISCHE WERKSTATTEN
1115 1116 1117 1118 1119
216-1280EA 106 EUR
216-1280 216-1281 216-1282 216-1283 216-1284
Dishes, evaporating
Haihdutusmaljat/Tarjottimet/Altaat Haihdutusaltaat/-maljat
Quartz glass, with spout.
  • High thermal shock resistance with low thermal expansion
  • High transmission from the UV to the IR spectrum range
  • High chemical resistance and purity
  • High electrical insulation properties
  • High radiation resistance
  • High temperature resistance and softening temperature
Sertifikaatit: DIN 12336