VWR®, Dropping Bottles, PTFE
Toimittaja: VWR Collection
216-0456EA 58.2 EUR
216-0456 216-0457
VWR®, Dropping Bottles, PTFE
Pullot Tippapullo
PTFE, white.
  • Flexible wall for easy use
  • Screw-fit cap
  • Guaranteed chemically resistant and inert
Suitable for dropping-out aggressive chemicals.