Pneumatic trough, pp, Nalgene®
Toimittaja: Thermo Fisher Scientific
6820-0010
216-0428EA 47.4 EUR
216-0428
Pneumatic trough, pp, Nalgene®
Opetusmateriaalit Physics Educational Materials Mechanics Learning Activities
Replace metal-type troughs with the polypropylene pneumatic trough.
  • Lightweight and rustproof
  • Gas tubing passes through a loop to prevent liquid from flowing into gas line
  • Liquid inlet and outlet connections accept ¼" int.Ø × ³/₈" Ø (6,4×9,5 mm) tubing
This trough features tapered sides for nesting and a built-in gas inlet. There are no sharp corners or separate parts to lose. The built-in platform accommodates four 250 ml bottles.