VWR®, Jäähdytyspakkaukset
Toimittaja: VWR Collection
216-0188EA 131 EUR
216-0188 216-0189 216-0190 216-0191 216-0192 216-0193 216-0194 216-0195 216-0196
VWR®, Jäähdytyspakkaukset
Kylmäpakkaukset
VWR® cooling bricks are available in foam brick or gel filled pouch formats. The semi-rigid foam brick will holds its shape when refrigerated or frozen. The gel filled pouch can be refrigerated or frozen to keep products cold or frozen.
  • Long lasting, high-performance temperature protection
  • Suitable for pharmaceuticals, clinical trials, diagnostic kits, chemicals and biologicals
  • Available in a range of sizes
These packs are designed to safeguard temperature-sensitive products during storage and transport. They can be chilled in a freezer, refrigerator,More Product Information