Pullo, leveä suuaukko, serie 303 brown
Toimittaja: Kautex
303-784583 303-784584 303-784585 303-784586 303-784587
215-9048EA 119 EUR
215-9048 215-9049 215-9050 215-9051 215-9052
Pullo, leveä suuaukko, serie 303 brown
Pullot Muovipullot
LDPE. Round. Brown colour. With closure.
  • For light sensitive materials
  • Protection against UV-light-transmission
  • Leakproof
  • Suitable for food
  • Wide neck opening means that the bottles are easy to fill and clean
  • Halal certified
Ideal for high-viscosity media, powders, granulates, and pastes.