Pullo, kapea suuaukko, laimennuspullo, kierrekorkilla, Nalgene®
Toimittaja: Thermo Fisher Scientific
2500-0280 2500-0380 2505-0380
215-7801EA 134 EUR
215-7801 215-7802 215-7812
Pullo, kapea suuaukko, laimennuspullo, kierrekorkilla, Nalgene®
Pullot Muovipullot
Polysulfoni (PSF), kirkas tai polypropeeni-kopolymeeri (PPCO), läpikuultava, PP-kierrekorkki.
  • PSF: Outstanding resistance to weak acids, bases and all aqueous solutions/PPCO: Outstanding resistance to strong acids, alcohols and bases
  • PSF: Temperature resistant from −100 to +165 °C/PPCO: Temperature resistant from −40 to +121 °C
  • PSF: Autoclavable, use with Tween shortens service life/PPCO: Autoclavable, sustains repeated cleaning and autoclaving with Tween
Compliant with the dilution methods described in the standard methods for waterMore Product Information