Tippapullo, lasia, hioksellisella pipetillä
Toimittaja: Kavalier
632425020050 632425020100 632425020250 632425023050 632425023100 632425023250
215-6246EA 38.5 EUR
215-6246 215-6247 215-6248 215-6265 215-6266 215-6267
Tippapullo, lasia, hioksellisella pipetillä
Pullot Tippapullo
Glass, clear or amber.
  • Good chemical and physical properties
  • For use in applications that do not involve high thermal stresses
  • Replaceable NS glass pipette
Amber to protect light-sensitive substances.