Bottles, narrow neck, LDPE, with thread
Toimittaja: KJS KUNSTSTOFFVERPACKUNGEN
E50 E100 E250 E500 E1000
215-6008EA 3.5 EUR
215-6008 215-6009 215-6010 215-6011 215-6012
Bottles, narrow neck, LDPE, with thread
Pullot Muovipullot
LDPE, natural colour.
  • Squeezable, so these flasks make excellent containers for droppers or wash bottles/sprayers
  • Please order caps (screw, dropper or spray caps) separately
Excellent for sampling, storing and transporting liquids.