High-temperature silicone cap liner, GL 45, PTFE coated, DURAN®
Toimittaja: DWK Life Sciences
292490909
215-4281EA 40.8 EUR
215-4281
High-temperature silicone cap liner, GL 45, PTFE coated, DURAN®
Septumi
Replacement GL 45 PTFE coated high-temperature silicone cap liner for the aluminium cap.