Jars, wide mouth, with screw and inner cap
Toimittaja: LAMAPLAST
22 23 53 24 26 25
215-4066EA 3.3 EUR
215-4066 215-4067 215-4070 215-4068 215-4069 215-4184
Jars, wide mouth, with screw and inner cap
Astiat ja purkit Astiat, yleiskäyttö
HDPE, translucent, with black screw cap.
  • Good chemical and thermal resistance
  • Wide neck for easy filling and cleaning
  • With screw cap and inner cap
  • For food use
Equipped with inner cap and screw cap, they can be easily usable in various fields : from chemical, to animal husbrandy and agriculture, until the use as containers for cosmetics and pharmaceuticals, and many others. Thanks to the wide mouth, they’re suitable for the storage of : creams, semi-solid substances, crystals, granulates and powders.