Multi-purpose cylinders, DURAN®
Toimittaja: DWK Life Sciences
213982101 213983406 213983603 213984608 213985201 213985304 213986806 213987408 213987708 213988001
215-1025EA 21.6 EUR
215-1025 215-3648 216-1850 215-3649 216-0273 215-3650 215-1022 216-0274 216-0275 215-3651
Multi-purpose cylinders, DURAN®
Mittalasit Säilytyssylinterit
DURAN®, borosilicate glass 3.3, clear, with round base.
  • Very good chemical resistance and high resistance to temperature change
  • No graduations
  • Rough ground rim without pouring lip