Kromatografian korkit, tulpat, kumia
Toimittaja: DWK Life Sciences
224100-178 224100-320 224100-172 224100-174 224100-175 224100-180 224100-072 W224100-081 224100-331 W224100-181 224100-160 224100-020 224100-060 224100-080 224100-170 W224100-342 W224100-173 224100-161
215-3487EA 1760 EUR
215-3487 215-4715 215-3484 215-3485 215-3486 215-4714 215-3481 215-4716 215-3490 215-4717 215-4713 215-4708 215-4709 215-4712 215-3482 215-4718 215-3492 548-2050
Kromatografian korkit, tulpat, kumia
Tulpat Kumi
Stoppers of various styles, sizes and rubber formulations.
  • Manufactured from the highest quality raw materials
  • Components manufactured to yield exceptional dimensional stability
  • Autoclavable
Straight plug stoppers provide maximum sealing for vials with straight-wall glass finish.
Snap-on style provides maximum sealing for vials with a blow-back glass finish.
Sleeve style stoppers feature a fold-over skirt for sealing in serum finish vials without a crimp closure.