Näytepullot, Tubular glass universal and bijou vials
Toimittaja: DWK Life Sciences
127028 127030 120128 128138
215-3392EA 204 EUR
215-3392 215-0095 215-3389 215-3393
Näytepullot, Tubular glass universal and bijou vials
Näytevialit ja pienet näyteastiat
These vials are suitable for media, diagnostic and sample collection applications. Type III, soda-lime glass, clear.
  • Good chemical resistance
  • For applications that do not require specific temperature resistance
  • PP or aluminium replacement screw caps optionally available (please enquire)