VWR® Sample Vials
Toimittaja: VWR Collection
215-1900EA 106 EUR
215-1900 215-1901
VWR® Sample Vials
Näytevialit ja pienet näyteastiat
These sample vials are made from PTFE.
  • Ideal for storing aggressive or expensive materials
  • Suitable for small scale reactions at low pressure
  • Tapered inner for easy removal of contents
  • Thick wall construction