Bottles, wide neck, PTFE, round, with screw cap
Toimittaja: Brand
130538 130548 130554 130562 130564
215-1744EA 166 EUR
215-1744 215-1746 215-1747 215-1748 215-1749
Bottles, wide neck, PTFE, round, with screw cap
Pullot Muovipullot
PTFE, strong walled.
  • Very good chemical resistance
  • Heat resistant from –200 up to +280 °C
  • Exceptionally smooth inner walls to prevent contamination
Robust, unbreakable construction. The special recessed grips on the cap and grooves in the flask make it easy to handle. Non stick, the wide mouth allows easy filling and cleaning.