Bottles, wide neck, square
Toimittaja: witeg Labortechnik
5.899.110 5.899.260 5.899.510
215-1674EA 15.4 EUR
215-1674 215-1675 215-1676
Bottles, wide neck, square
Pullot Lasipullot
Soda-lime glass, coated.
  • With PU coating to protect the bottle from scratches, splinters, and leaking liquids
Without cap and pouring ring.