Pullo, kapea suuaukko, normaalilla hiostulpalla, DURAN®
Toimittaja: DWK Life Sciences
231652408 231653601 231654409 231655405 231681706 231682402 231683604 231684403 231685408
215-1482EA 12.3 EUR
215-1482 215-1282 215-1016 215-1483 215-1484 215-1021 215-1020 215-1735 215-1485
Pullo, kapea suuaukko, normaalilla hiostulpalla, DURAN®
Pullot Lasipullot
Soda-lime glass, clear or brown, with standard ground joint and glass stopper.
  • Good chemical and physical properties
  • For use in applications that are not associated with high thermal loads
  • Stable and tough
Available in crystal clear glass or brown tinted to protect light-sensitive substances.
Sertifikaatit: DIN 12036 ja ISO 4796