Pullo, leveä suuaukko, with flared rim, DURAN®
Toimittaja: DWK Life Sciences
211812408 211813601 211814409 211815405 211816307 211817303 211818608 211819107
215-1374EA 16.4 EUR
215-1374 215-1419 215-1376 215-1420 215-1421 215-1422 215-1380 215-1381
Pullo, leveä suuaukko, with flared rim, DURAN®
Pullot Lasipullot
DURAN®, borosilicate glass 3.3, clear, ungrounded neck.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
  • Glass Type I according to USP, EP and JP
Round, with printed volumes and white labelling area.
Sertifikaatit: DIN 12038.