Pullo, kapea suuaukko, pakkauspullo, kierrekorkilla, Nalgene®
Toimittaja: Thermo Fisher Scientific
312085-0001 312089-0001 312089-0004 312089-0032 312085-0002 312085-0004 312085-0008 312085-0016 312085-0032 312089-0002 312089-0008 312089-0016
215-1007EA 1150 EUR
215-1007 215-1009 215-1017 215-1432 215-1410 215-1411 215-1412 215-1414 215-1415 215-1416 215-1430 215-1431
Pullo, kapea suuaukko, pakkauspullo, kierrekorkilla, Nalgene®
Pullot Muovipullot
HDPE, translucent or amber with PP screw cap.
  • Good chemical resistance
  • Heat resistant from –100 to +120 °C
  • Leakproof
Economical, robust bottles for storing and transporting powders, liquids or pastes. Amber bottles to protect light-sensitive substances.
Varoitus: The product(s) on this page are outside the scope of the EU Directive 2019/904.
Order Now
TEKNISET TIEDOT
Environmentally Preferable Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact
Ympäristösertifikaatti NAR Certificate of Retirement NAR Certificate of Retirement IRECS; ISO 14001 (Certificate Registration No. EMS 752603) NAR Certificate of Retirement NAR Certificate of Retirement IRECS; ISO 14001 (Certificate Registration No. EMS 752603) NAR Certificate of Retirement NAR Certificate of Retirement NAR Certificate of Retirement NAR Certificate of Retirement NAR Certificate of Retirement NAR Certificate of Retirement