Pullo, leveä suuaukko, normaalhios, hiostulpalla, DURAN®
Toimittaja: DWK Life Sciences
211852403 211853605 211854404 211855409 211856302 211857307 211858603 211859102 211881701 211882406 211883608 211884407 211885403 211886305 211887301 211888606 211889105
215-1314EA 58.8 EUR
215-1314 215-1315 215-1316 215-1317 215-1318 215-1319 215-1320 215-1321 215-1343 215-1344 215-1345 215-1346 215-1347 215-1348 215-1349 215-1350 215-1351
Pullo, leveä suuaukko, normaalhios, hiostulpalla, DURAN®
Pullot Lasipullot
DURAN®, borosilicate glass 3.3, clear or amber, with standard ground joint and glass stopper.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
  • Glass Type I according to USP, EP and JP
  • Amber glass meets USP 671 and Eur. Ph. 3.2.1. for protection against UV light
Marked with volume and equipped with a white labelling field. Amber to protect light-sensitive materials.
With printed retrace code (batch identification).
ISO 4796-2
Sertifikaatit: ISO 4796-2.