Holders for round bottom flasks, Flaskup™, SP Bel-Art
Toimittaja: Bel-Art Products, a Part of SP
F389512004 F389512006 F389512010 F389512012
214-1952EA 74.8 EUR
214-1952 214-1953 214-1954 214-1955
Holders for round bottom flasks, Flaskup™, SP Bel-Art
Pullotuet Pullotuet
Holders made from PP for round bottom flasks.
  • Inner wall has a slight taper to the open bottom
  • Allows variety of flask shapes and sizes to fit into one holder
  • Wide base keeps flasks stable
  • Side slots can be used as sight windows
  • Temperature resistant upto 121 °C
Tilaustiedot: Available as 3 per package, 8 packages per case and assortment packs.