VWR®, PC Erlenmeyer Shaker Flasks, Baffled Bottom
Toimittaja: VWR Collection
214-0458EA 160 EUR
214-0458 214-0459 214-0460 214-0461 214-0462 214-0463 214-0464 214-0465
VWR®, PC Erlenmeyer Shaker Flasks, Baffled Bottom
Kolvit ja erlenmeyerit Erlenmeyerkolvit
VWR® PC Erlenmeyer shaker flasks provide excellent optical clarity and mechanical strength. They are ideal for shaker culture applications and storage. The baffled bottom improves mixing and increase gas exchange.
  • Temperature range: –80 to +121 °C
  • Unlike glass, flasks will not break when dropped
  • Moulded-in graduations for accuracy
  • Vent cap option for continuous gas exchange while ensuring sterility and preventing leakage
  • Non cytotoxic, DNase-/RNase-free and non-pyrogenic
  • Unique baffled design with a moulded-in '1/3 Fill' line for customer convenience