Dekantteri, matala malli, PTFE
Toimittaja: Brand
90320 90328 90338 90344 90348 90352 90354 90362 90364
213-1554EA 34.1 EUR
213-1554 213-1556 213-1558 213-1559 213-1560 213-1561 213-1562 213-1564 213-1565
Dekantteri, matala malli, PTFE
Dekantterit
PTFE, with spout.
  • Excellent chemical resistance
  • Temperature resistant from −200 to +280 °C
  • Flat bottom for good heat transfer, for example when used on regulated hotplates
Thick walled, with reinforced lip and exceptionally smooth inner walls to prevent contamination. Non stick. Low gas permeability.