Beakers, low or tall form
Toimittaja: Kavalier
N632427150005 N632427150010 N632427150025 N632427150150 632417010250 632417010400 632417010600 632417010800 632417010940 632411012050 N632427152100 N632427152150 632417012400 632417012600 632417012800 632411010966 632417010950 632417010952 632417010956 632417012250 632417012940 632417012950
213-1041EA 63.4 EUR
213-1041 213-1042 213-1043 213-1046 213-1047 213-1048 213-1049 213-1050 213-1051 213-1061 213-1062 213-1063 213-1065 213-1066 213-1067 KAVA632411010966 KAVA632417010950 KAVA632417010952 KAVA632417010956 KAVA632417012250 KAVA632417012940 KAVA632417012950
Beakers, low or tall form
Dekantterit
Borosilicate glass, grade 3,3.
  • Excellent chemical resistance
  • High temperature resistance
  • Autoclavable
Ideal for general laboratory use. With spout and white graduations.
Sertifikaatit: DIN 12331 (exept 25 ml/5000 ml tall form and 10000 ml low form supplementary to DIN). ISO 3819.