Measuring beaker with handle, Azlon®
Toimittaja: SciLabware
JPM0250P JPM0500P JPM1000P JPM2000P JPM3000P JPM5000P JPM0100P
213-0154EA 3.5 EUR
213-0154 213-0155 213-0156 213-0157 213-0158 213-0159 213-0153
Measuring beaker with handle, Azlon®
Dekantterit
PP, läpikuultava.
  • Good chemical resistance
  • Resistant to heat up to 121 °C
  • Smooth surfaces for easy, fast cleaning
Ideal for general laboratory use and a wide variety of other applications. Clear moulded graduations. Ergonomically designed handle with thumb grip for easy carrying.