Dekantteri, matala malli, Azlon®
Toimittaja: SciLabware
BPM0050P BPM0100P BPM0500P BPM1000P BPM2000P BPM0250P
213-0107EA 23.1 EUR
213-0107 213-0108 213-0110 213-0112 213-0113 213-0109
Dekantteri, matala malli, Azlon®
Dekantterit
PP, läpikuultava.
  • Tapered stacking style - space saving and easy to handle
  • Autoclavable, durable with good chemical resistance
  • Precise non-drip pouring spouts, with easy-to-read moulded graduations
Sertifikaatit: Comply with ISO 7056.