Dekantteri, matala malli
Toimittaja: SciLabware
1000/03M 1000/30D 1000/34D 1000/38D 1000/11M 1000/02M 1000/04D 1000/06D 1000/10D 1000/14D 1000/18D 1000/20D 1000/22D 1000/28D 1000/05M
213-0010EA 90.8 EUR
213-0010 213-0268 213-0022 213-0023 213-0015 213-0009 213-0011 213-0013 213-0014 213-0016 213-0017 213-0018 213-0019 213-0020 213-0012
Dekantteri, matala malli
Dekantterit
PYREX® borosilicate glass, clear, with spout.
  • Very good chemical resistance
  • High temperature resistance
  • Autoclavable
Ideal for general laboratory use. With spout, white graduations and labelling area.
Sertifikaatit: ISO 3819 and DIN 12331 - Beaker 10000 ml supplementary to ISO range and graduations are approximate.