Test tube racks
Toimittaja: DELTALAB
Z-100 Z-250 Z-300 Z-050 Z-075 Z-500 Z-600 Z-800
212-8401EA 29.9 EUR
212-8401 212-8402 212-8403 212-8422 212-8423 212-8801 212-8803 212-8823
Test tube racks
Koeputkitelineet Tube Racks
Test tube racks, made of aluminium or stainless steel.
  • Stackable
  • Alpha-numerical index for better sample location
  • Good chemical and temperature resistance
Toimitustiedot: Supplied encased and interlocked, wrapped in a protecting film.